(1R,6S)-6-Amino-2,2-difluorocyclohexanol 95%

Supplier: Apollo Scientific

PC520830-250MG PC520830-500MG
APOSPC520830-250MGEA 792 EUR
APOSPC520830-250MG APOSPC520830-500MG
(1R,6S)-6-Amino-2,2-difluorocyclohexanol 95%
(1R,6S)-6-Amino-2,2-difluorocyclohexanol
Formula: C₆H₁₁F₂NO
MW: 151,154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28118180
CAS Number: 1374973-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR