3-Amino-4-hydroxyphenylsulphur pentafluoride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-4-(pentafluorosulphanyl)phenol, 2-Hydroxy-5-(pentafluorothio)aniline
PC5209-250MG
PC5209-1G
APOSPC5209-250MGEA
263
EUR
APOSPC5209-250MG
APOSPC5209-1G
3-Amino-4-hydroxyphenylsulphur pentafluoride
3-Amino-4-hydroxyphenylsulphur pentafluoride
Formula:
C₆H₆F₅NOS MW: 235,175 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11501041 CAS Number: 1159512-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...