3-Fluoropropan-1-amine
Supplier: Apollo Scientific
|
Danger
|
PC52117-100MG
PC52117-1G
PC52117-250MG
APOSPC52117-100MGEA
190
EUR
APOSPC52117-100MG
APOSPC52117-1G
APOSPC52117-250MG
3-Fluoropropan-1-amine
3-Fluoropropan-1-amine
Formula:
C₃H₈FN MW: 77,101 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01694865 CAS Number: 462-41-9 UN: 2733 ADR: 3+8,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...