2-Bromo-3,5-difluorobenzyl fluoride 95%
Supplier: Apollo Scientific
|
Danger
|
PC52169-1G
PC52169-250MG
APOSPC52169-1GEA
69.6
EUR
APOSPC52169-1G
APOSPC52169-250MG
2-Bromo-3,5-difluorobenzyl fluoride 95%
2-Bromo-3,5-difluorobenzyl fluoride
Formula:
C₇H₄BrF₃ MW: 225.006 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31722921 CAS Number: 2366994-41-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...