2-Bromo-3,6-difluorobenzal fluoride

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-3-(difluoromethyl)-1,4-difluorobenzene

PC52176-1G PC52176-250MG PC52176-5G
APOSPC52176-1GEA 105 EUR
APOSPC52176-1G APOSPC52176-250MG APOSPC52176-5G
2-Bromo-3,6-difluorobenzal fluoride
2-Bromo-3,6-difluorobenzal fluoride
Formula: C₇H₃BrF₄
MW: 242,996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28166242
CAS Number: 1672663-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR