3-Bromo-2,4-difluorobenzal fluoride

Supplier: Apollo Scientific
Warning

PC52178-1G PC52178-250MG PC52178-5G
APOSPC52178-1GEA 522 EUR
APOSPC52178-1G APOSPC52178-250MG APOSPC52178-5G
3-Bromo-2,4-difluorobenzal fluoride
3-Bromo-2,4-difluorobenzal fluoride
Formula: C₇H₃BrF₄
MW: 242,996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737886
CAS Number: 1804408-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR