4-Bromo-3-(trifluoromethoxy)benzal fluoride
Supplier: Apollo Scientific
|
Warning
|
PC52182-1G
PC52182-250MG
PC52182-5G
APOSPC52182-1GEA
575
EUR
APOSPC52182-1G
APOSPC52182-250MG
APOSPC52182-5G
4-Bromo-3-(trifluoromethoxy)benzal fluoride
4-Bromo-3-(trifluoromethoxy)benzal fluoride
Formula:
C₈H₄BrF₅O MW: 291.013 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31812119 CAS Number: 2167351-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...