4-Bromo-2-chlorobenzal fluoride

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-chloro-1-(difluoromethyl)benzene

PC52183-1G PC52183-250MG PC52183-5G PC52183-10G PC52183-25G
APOSPC52183-1GEA 522 EUR
APOSPC52183-1G APOSPC52183-250MG APOSPC52183-5G APOSPC52183-10G APOSPC52183-25G
4-Bromo-2-chlorobenzal fluoride
4-Bromo-2-chlorobenzal fluoride
Formula: C₇H₄BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD18391084
CAS Number: 1254034-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR