5-Bromo-2-chlorobenzal fluoride

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-1-chloro-2-(difluoromethyl)benzene

PC52184-1G PC52184-250MG
APOSPC52184-1GEA 522 EUR
APOSPC52184-1G APOSPC52184-250MG
5-Bromo-2-chlorobenzal fluoride
5-Bromo-2-chlorobenzal fluoride
Formula: C₇H₄BrClF₂
MW: 241.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698636
CAS Number: 627527-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR