4-(4-Bromo-3-fluoro-phenyl)-1H-1,2,4-triazol-5-one

Supplier: Apollo Scientific
Warning

PC52210-1G PC52210-250MG
APOSPC52210-1GEA 576 EUR
APOSPC52210-1G APOSPC52210-250MG
4-(4-Bromo-3-fluoro-phenyl)-1H-1,2,4-triazol-5-one
4-(4-Bromo-3-fluoro-phenyl)-1H-1,2,4-triazol-5-one
Formula: C₈H₅BrFN₃O
MW: 258,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30310500
CAS Number: 1696800-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR