4-(Difluoromethyl)benzylamine

Supplier: Apollo Scientific
Warning

Synonyms: (4-(Difluoromethyl)phenyl)methanamine

PC52214-1G PC52214-250MG
APOSPC52214-1GEA 430 EUR
APOSPC52214-1G APOSPC52214-250MG
4-(Difluoromethyl)benzylamine
4-(Difluoromethyl)benzylamine
Formula: C₈H₉F₂N
MW: 157,161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18791602
CAS Number: 754920-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR