5-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-indole, Technical Grade

Supplier: Apollo Scientific

PC52230-1G PC52230-250MG
APOSPC52230-1GEA 654 EUR
APOSPC52230-1G APOSPC52230-250MG
5-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-indole, Technical Grade
5-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-indole
Formula: C₁₀H₅F₆NO
MW: 269,143 g/mol
Storage Temperature: Freezer
MDL Number: MFCD20226849
CAS Number: 1007235-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR