(2S)-2-Amino-4,4,4-trifluorobutanoic acid, N-BOC protected

Supplier: Apollo Scientific
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-4,4,4-trifluorobutanoic acid, L-2-[(tert-Butoxycarbonyl)amino]-4,4,4-trifluorobutyric acid, (2S)-2-[(tert-Butoxycarbonyl)amino]-4,4,4-trifluorobutanoic acid

PC5241-250MG PC5241-100MG
APOSPC5241-250MGEA 224 EUR
APOSPC5241-250MG APOSPC5241-100MG
(2S)-2-Amino-4,4,4-trifluorobutanoic acid, N-BOC protected
(2S)-2-Amino-4,4,4-trifluorobutanoic acid, N-BOC protected
Formula: C₉H₁₄F₃NO₄
MW: 257.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08532484
CAS Number: 181128-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR