2,6-Dichloro-4-(trifluoromethoxy)benzamide 95%
Supplier: Apollo Scientific
PC52459-1G
PC52459-250MG
APOSPC52459-1GEA
524
EUR
APOSPC52459-1G
APOSPC52459-250MG
2,6-Dichloro-4-(trifluoromethoxy)benzamide 95%
2,6-Dichloro-4-(trifluoromethoxy)benzamide
Formula:
C₈H₄Cl₂F₃NO₂ MW: 274,024 g/mol Storage Temperature: Ambient |
CAS Number:
886503-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...