2-(Difluoromethoxy)cinnamic acid 95%

Supplier: Apollo Scientific

PC52467-1G PC52467-5G
APOSPC52467-1GEA 141 EUR
APOSPC52467-1G APOSPC52467-5G
2-(Difluoromethoxy)cinnamic acid 95%
2-(Difluoromethoxy)cinnamic acid
Formula: C₁₀H₈F₂O₃
MW: 214,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419433
CAS Number: 884-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR