3-[2-(Difluoromethoxy)phenyl]propionic acid 95%
Supplier: Apollo Scientific
PC52473-1G
PC52473-250MG
PC52473-5G
APOSPC52473-1GEA
524
EUR
APOSPC52473-1G
APOSPC52473-250MG
APOSPC52473-5G
3-[2-(Difluoromethoxy)phenyl]propionic acid 95%
3-[2-(Difluoromethoxy)phenyl]propionic acid
Formula:
C₁₀H₁₀F₂O₃ MW: 216,181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09951709 CAS Number: 1017030-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...