2-Ethylbenzotrifluoride 98%
Supplier: Apollo Scientific
Synonyms:
1-Ethyl-2-(trifluoromethyl)benzene
PC52482-1G
APOSPC52482-1GEA
132
EUR
APOSPC52482-1G
2-Ethylbenzotrifluoride 98%
2-Ethylbenzotrifluoride
Formula:
C₉H₉F₃ MW: 174,163 g/mol Storage Temperature: Ambient |
CAS Number:
74423-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...