3,3,4,4,5,5,6,6,7,7,7-Undecafluoroheptan-2-ol

Supplier: Apollo Scientific
Warning

PC5261-1G PC5261-250MG
APOSPC5261-1GEA 959 EUR
APOSPC5261-1G APOSPC5261-250MG
3,3,4,4,5,5,6,6,7,7,7-Undecafluoroheptan-2-ol
3,3,4,4,5,5,6,6,7,7,7-Undecafluoroheptan-2-ol
Formula: C₇H₅F₁₁O
Storage Temperature: Ambient
MDL Number: MFCD08460482
CAS Number: 914637-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR