b-Amino-3-fluorobenzenepropanol 97%

Supplier: Apollo Scientific

PC530044-1G PC530044-250MG PC530044-500MG
APOSPC530044-1GEA 783 EUR
APOSPC530044-1G APOSPC530044-250MG APOSPC530044-500MG
b-Amino-3-fluorobenzenepropanol 97%
b-Amino-3-fluorobenzenepropanol
Formula: C₉H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD09744986
CAS Number: 35373-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR