1-(2,4-Difluorophenyl)ethanamine 97%
Supplier: Apollo Scientific
PC530059-1G
PC530059-500MG
APOSPC530059-1GEA
389
EUR
APOSPC530059-1G
APOSPC530059-500MG
1-(2,4-Difluorophenyl)ethanamine 97%
1-(2,4-Difluorophenyl)ethanamine
Formula:
C₈H₉F₂N MW: 157,161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116332 CAS Number: 603951-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...