2-(Trifluoromethoxy)phenylacetylene 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Ethynylphenyl trifluoromethyl ether, 1-Ethynyl-2-(trifluoromethoxy)benzene

PC53008-5G PC53008-1G PC53008-250MG
APOSPC53008-5GEA 219 EUR
APOSPC53008-5G APOSPC53008-1G APOSPC53008-250MG
2-(Trifluoromethoxy)phenylacetylene 95+%
2-(Trifluoromethoxy)phenylacetylene
Formula: C₉H₅F₃O
MW: 186.131 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03839994
CAS Number: 886363-40-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR