3-Bromo-2-chloro-6-fluoroiodobenzene

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-2-chloro-4-fluoro-3-iodobenzene

PC53018-1G PC53018-250MG PC53018-5G
APOSPC53018-1GEA 290 EUR
APOSPC53018-1G APOSPC53018-250MG APOSPC53018-5G
3-Bromo-2-chloro-6-fluoroiodobenzene
3-Bromo-2-chloro-6-fluoroiodobenzene
Formula: C₆H₂BrClFI
MW: 335,34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28140548
CAS Number: 1820649-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR