3'-Bromo-2'-chloro-6'-fluoroacetophenone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(3-Bromo-2-chloro-6-fluorophenyl)ethanone

PC53027-1G PC53027-5G PC53027-250MG
APOSPC53027-1GEA 488 EUR
APOSPC53027-1G APOSPC53027-5G APOSPC53027-250MG
3'-Bromo-2'-chloro-6'-fluoroacetophenone
3'-Bromo-2'-chloro-6'-fluoroacetophenone
Formula: C₈H₅BrClFO
MW: 251,48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28140553
CAS Number: 1704367-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR