2-Fluoro-3-nitrobenzonitrile
Supplier: Apollo Scientific
|
Warning
|
PC53131-1G
PC53131-5G
PC53131-25G
APOSPC53131-1GEA
72.7
EUR
APOSPC53131-1G
APOSPC53131-5G
APOSPC53131-25G
2-Fluoro-3-nitrobenzonitrile
2-Fluoro-3-nitrobenzonitrile
Formula:
C₇H₃FN₂O₂ MW: 166,109 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11849938 CAS Number: 1214328-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...