2-Fluoro-5-(hydroxymethyl)phenol 95+%

Supplier: Apollo Scientific

Synonyms: 4-Fluoro-3-hydroxybenzyl alcohol, 4-Fluoro-4-hydroxybenzyl alcohol

PC53135-1G PC53135-250MG PC53135-5G
APOSPC53135-1GEA 147 EUR
APOSPC53135-1G APOSPC53135-250MG APOSPC53135-5G
2-Fluoro-5-(hydroxymethyl)phenol 95+%
2-Fluoro-5-(hydroxymethyl)phenol
Formula: C₇H₇FO₂
MW: 142,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031832
CAS Number: 934241-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR