2-Bromo-5-chloro-4-fluorobenzaldehyde

Supplier: Apollo Scientific

PC53327-1G PC53327-250MG PC53327-5G
APOSPC53327-1GEA 412 EUR
APOSPC53327-1G APOSPC53327-250MG APOSPC53327-5G
2-Bromo-5-chloro-4-fluorobenzaldehyde
2-Bromo-5-chloro-4-fluorobenzaldehyde
Formula: C₇H₃BrClFO
MW: 237,454 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28733864
CAS Number: 1806838-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR