Ethyl 2-bromo-5-fluorobenzoate 97%

Supplier: Apollo Scientific
Warning

PC53349-25G
APOSPC53349-25GEA 75.4 EUR
APOSPC53349-25G
Ethyl 2-bromo-5-fluorobenzoate 97%
Ethyl 2-bromo-5-fluorobenzoate
Formula: C₉H₈BrFO₂
MW: 247,061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204688
CAS Number: 139911-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR