Methyl 4-bromo-2-fluoro-5-nitrobenzoate

Supplier: Apollo Scientific
Warning

PC53375-1G PC53375-250MG PC53375-5G
APOSPC53375-1GEA 224 EUR
APOSPC53375-1G APOSPC53375-250MG APOSPC53375-5G
Methyl 4-bromo-2-fluoro-5-nitrobenzoate
Methyl 4-bromo-2-fluoro-5-nitrobenzoate
Formula: C₈H₅BrFNO₄
MW: 278,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12173001
CAS Number: 1153285-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR