4-Bromo-2-fluoro-5-nitrobenzyl alcohol
Supplier: Apollo Scientific
|
Warning
|
PC53376-1G
PC53376-250MG
APOSPC53376-1GEA
1320
EUR
APOSPC53376-1G
APOSPC53376-250MG
4-Bromo-2-fluoro-5-nitrobenzyl alcohol
4-Bromo-2-fluoro-5-nitrobenzyl alcohol
Formula:
C₇H₅BrFNO₃ MW: 250,022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17168699 CAS Number: 1379332-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...