4-Bromo-2-fluoro-5-iodobenzoic acid

Supplier: Apollo Scientific
Warning

PC53406-1G PC53406-250MG PC53406-5G
APOSPC53406-1GEA 473 EUR
APOSPC53406-1G APOSPC53406-250MG APOSPC53406-5G
4-Bromo-2-fluoro-5-iodobenzoic acid
4-Bromo-2-fluoro-5-iodobenzoic acid
Formula: C₇H₃BrFIO₂
MW: 344.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107175
CAS Number: 1521585-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR