8-Bromo-6-(trifluoromethoxy)-1,2,3,4-tetrahydroquinoline-2-one 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
8-Bromo-6-(trifluoromethoxy)-1,2,3,4-tetrahydroquinoline-2-one
PC53416-1G
PC53416-250MG
APOSPC53416-1GEA
609
EUR
APOSPC53416-1G
APOSPC53416-250MG
8-Bromo-6-(trifluoromethoxy)-1,2,3,4-tetrahydroquinoline-2-one 95+%
8-Bromo-6-(trifluoromethoxy)-1,2,3,4-tetrahydroquinoline-2-one
Formula:
C₁₀H₇BrF₃NO₂ MW: 310.067 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30474684 CAS Number: 2149597-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...