3-Fluoro-5-nitrophenol
Supplier: Apollo Scientific
|
Warning
|
PC53464-1G
PC53464-250MG
APOSPC53464-1GEA
288
EUR
APOSPC53464-1G
APOSPC53464-250MG
3-Fluoro-5-nitrophenol
3-Fluoro-5-nitrophenol
Formula:
C₆H₄FNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD09832852 CAS Number: 2369-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...