3-(Trifluoromethylsulphonyl)benzoic acid

Supplier: Apollo Scientific
Warning

PC53480-1G PC53480-250MG
APOSPC53480-1GEA 412 EUR
APOSPC53480-1G APOSPC53480-250MG
3-(Trifluoromethylsulphonyl)benzoic acid
3-(Trifluoromethylsulphonyl)benzoic acid
Formula: C₈H₅F₃O₄S
MW: 254,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00766763
CAS Number: 952-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR