5-Bromo-4-fluoro-2-methoxybenzoic acid 97%

Supplier: Apollo Scientific
Warning

PC535006-100MG PC535006-250MG
APOSPC535006-100MGEA 184 EUR
APOSPC535006-100MG APOSPC535006-250MG
5-Bromo-4-fluoro-2-methoxybenzoic acid 97%
5-Bromo-4-fluoro-2-methoxybenzoic acid
Formula: C₈H₆BrFO₃
MW: 249,034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527529
CAS Number: 95383-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR