Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%

Supplier: Apollo Scientific
Warning

PC535008-100MG PC535008-250MG
APOSPC535008-100MGEA 50.9 EUR
APOSPC535008-100MG APOSPC535008-250MG
Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%
Methyl 4-bromo-2-fluoro-6-methoxybenzoate
Formula: C₉H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD23707517
CAS Number: 1427415-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR