2-(4-Acetyl-2-fluorophenyl)acetic acid 95%

Supplier: Apollo Scientific
Warning

PC535018-1G PC535018-250MG
APOSPC535018-1GEA 159 EUR
APOSPC535018-1G APOSPC535018-250MG
2-(4-Acetyl-2-fluorophenyl)acetic acid 95%
2-(4-Acetyl-2-fluorophenyl)acetic acid
Formula: C₁₀H₉FO₃
MW: 196,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30188795
CAS Number: 2007925-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR