2-Chloro-6-fluoronicotinic acid 95+%

Supplier: Apollo Scientific
Warning

PC535024-100MG PC535024-250MG
APOSPC535024-100MGEA 98.6 EUR
APOSPC535024-100MG APOSPC535024-250MG
2-Chloro-6-fluoronicotinic acid 95+%
2-Chloro-6-fluoronicotinic acid
Formula: C₆H₃ClFNO₂
MW: 175,545 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185820
CAS Number: 1211533-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR