6-Bromo-5-fluoronicotinaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
PC535061-100MG
PC535061-250MG
APOSPC535061-100MGEA
45.1
EUR
APOSPC535061-100MG
APOSPC535061-250MG
6-Bromo-5-fluoronicotinaldehyde 97%
6-Bromo-5-fluoronicotinaldehyde
Formula:
C₆H₃BrFNO MW: 203.997 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16606505 CAS Number: 1227588-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...