7,8-Difluorochroman-4-amine 95+%

Supplier: Apollo Scientific
Warning

PC535083-100MG PC535083-250MG PC535083-1G
APOSPC535083-100MGEA 58 EUR
APOSPC535083-100MG APOSPC535083-250MG APOSPC535083-1G
7,8-Difluorochroman-4-amine 95+%
7,8-Difluorochroman-4-amine
Formula: C₉H₉F₂NO
MW: 185,171 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07374042
CAS Number: 886762-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR