6-Fluoro-2,2'-bipyridine 95+%

Supplier: Apollo Scientific
Warning

PC535084-250MG PC535084-1G
APOSPC535084-250MGEA 98.6 EUR
APOSPC535084-250MG APOSPC535084-1G
6-Fluoro-2,2'-bipyridine 95+%
6-Fluoro-2,2'-bipyridine
Formula: C₁₀H₇FN₂
MW: 174,174 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22576117
CAS Number: 1223063-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR