1-Fluoro-3-(2-isocyanatovinyl)benzene 95%
Supplier: Apollo Scientific
|
Warning
|
PC53526-250MG
PC53526-100MG
PC53526-25MG
APOSPC53526-250MGEA
998
EUR
APOSPC53526-250MG
APOSPC53526-100MG
APOSPC53526-25MG
1-Fluoro-3-(2-isocyanatovinyl)benzene 95%
1-Fluoro-3-(2-isocyanatovinyl)benzene
When melted, may contain small amount of sediment
Formula:
C₉H₆FNO Storage Temperature: Ambient |
MDL Number:
MFCD21086110 CAS Number: 2044498-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...