1-(4-Bromo-2-fluoro-phenyl)imidazolidin-2-one

Supplier: Apollo Scientific
Warning

PC53591-1G PC53591-250MG
APOSPC53591-1GEA 354 EUR
APOSPC53591-1G APOSPC53591-250MG
1-(4-Bromo-2-fluoro-phenyl)imidazolidin-2-one
1-(4-Bromo-2-fluoro-phenyl)imidazolidin-2-one
Formula: C₉H₈BrFN₂O
Storage Temperature: Ambient
MDL Number: MFCD24133750
CAS Number: 160132-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR