1-(4-Bromo-3-fluoro-phenyl)imidazolidin-2-one
Supplier: Apollo Scientific
|
Warning
|
PC53592-1G
PC53592-250MG
APOSPC53592-1GEA
415
EUR
APOSPC53592-1G
APOSPC53592-250MG
1-(4-Bromo-3-fluoro-phenyl)imidazolidin-2-one
1-(4-Bromo-3-fluoro-phenyl)imidazolidin-2-one
Formula:
C₉H₈BrFN₂O MW: 259,075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24134033 CAS Number: 1503577-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...