1,2,4,4,4-Pentachloropentafluorobutane 96%
Supplier: Apollo Scientific
PC5369F-5G
PC5369F-1G
APOSPC5369F-5GEA
477
EUR
APOSPC5369F-5G
APOSPC5369F-1G
1,2,4,4,4-Pentachloropentafluorobutane 96%
1,2,4,4,4-Pentachloropentafluorobutane
Formula:
C₄Cl₅F₅ Boiling Pt: 163…171 °C Density: 1,788 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00236124 CAS Number: 375-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...