2-Chloro-4-fluoro-5-methoxy-phenylamine > 95%
Supplier: Apollo Scientific
PC540006-1G
PC540006-250MG
APOSPC540006-1GEA
219
EUR
APOSPC540006-1G
APOSPC540006-250MG
2-Chloro-4-fluoro-5-methoxy-phenylamine > 95%
2-Chloro-4-fluoro-5-methoxy-phenylamine
Formula:
C₇H₇ClFNO MW: 175,588 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20696563 CAS Number: 98404-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...