3,3,4,4,5,5,5-Heptafluoropentan-1-ol
Supplier: Apollo Scientific
|
Warning
|
PC5434-1G
PC5434-25G
PC5434-5G
APOSPC5434-1GEA
79.9
EUR
APOSPC5434-1G
APOSPC5434-25G
APOSPC5434-5G
3,3,4,4,5,5,5-Heptafluoropentan-1-ol
3,3,4,4,5,5,5-Heptafluoropentan-1-ol
Formula:
C₅H₅F₇O Boiling Pt: 116…117 °C (630 mm Hg) Density: 1,506 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD04039292 CAS Number: 755-40-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...