3-Bromo-6-fluoro-1H-indazole
Supplier: Apollo Scientific
|
Warning
|
PC5509-1G
PC5509-250MG
APOSPC5509-1GEA
109
EUR
APOSPC5509-1G
APOSPC5509-250MG
3-Bromo-6-fluoro-1H-indazole
3-Bromo-6-fluoro-1H-indazole
Formula:
C₇H₄BrFN₂ MW: 215,022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781603 CAS Number: 885522-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...