Lithium difluoroacetate

Supplier: Apollo Scientific
Warning

PC5514-1G PC5514-5G
APOSPC5514-1GEA 102 EUR
APOSPC5514-1G APOSPC5514-5G
Lithium difluoroacetate
Lithium difluoroacetate
Formula: C₂HF₂O₂·Li
Storage Temperature: Ambient
MDL Number: MFCD03094294
CAS Number: 74956-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR