Fluorad® FC-100

Supplier: Apollo Scientific
Danger

PC5533-2.5KG PC5533-10KG
APOSPC5533-2.5KGEA 136 EUR
APOSPC5533-2.5KG APOSPC5533-10KG
Fluorad® FC-100
Fluorad® FC-100
Boiling Pt: 98 °C
Density: 1,1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03099297
CAS Number: 147335-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR