5-Bromo-3-hydroxy-2-nitrobenzotrifluoride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-2-nitro-3-(trifluoromethyl)phenol
PC56047-250MG
APOSPC56047-250MGEA
21.9
EUR
APOSPC56047-250MG
5-Bromo-3-hydroxy-2-nitrobenzotrifluoride
5-Bromo-3-hydroxy-2-nitrobenzotrifluoride
Formula:
C₇H₃BrF₃NO₃ MW: 286,003 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30746953 CAS Number: 2092867-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...